Skip to Content
Merck
All Photos(1)

Key Documents

46065

Sigma-Aldrich

Ethidium bromide

for fluorescence, ≥95.0% (HPLC)

Synonym(s):

3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, EtBr, Homidium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20BrN3
CAS Number:
Molecular Weight:
394.31
Beilstein:
3642536
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for fluorescence

Assay

≥95.0% (HPLC)

mp

260-262 °C (dec.) (lit.)

solubility

H2O: 10 mg/mL, opaque, very dark red

fluorescence

λex 480 nm; λem 620 nm in H2O
λex 530 nm; λem 600 nm in 50 mM phosphate buffer pH 7.0 (upon binding to DNA)

SMILES string

[Br-].CC[n+]1c(-c2ccccc2)c3cc(N)ccc3c4ccc(N)cc14

InChI

1S/C21H19N3.BrH/c1-2-24-20-13-16(23)9-11-18(20)17-10-8-15(22)12-19(17)21(24)14-6-4-3-5-7-14;/h3-13,23H,2,22H2,1H3;1H

InChI key

ZMMJGEGLRURXTF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethidium bromide (EtBr) undergoes complexation with nucleic acids (DNA and RNA) specifically to the saturated sites. These complexes decomposes by employing a cation-exchange resin. It has been reported to inhibit the production of mitochondrial RNA in HeLa cells. On interaction with helical polynucleotides under high salt medium, it forms fluorescent complex (having high fluorescence quantum yield).

Application

Ethidium bromide (EtBr) is the most commonly used nucleic acid stain for PAGE or agarose gel electrophoresis. The fluorescence of EtBr increases 21-fold upon binding to double-stranded RNA and 25-fold on binding double-stranded DNA so that destaining the background is not necessary with a low stain concentration (10 μg/ml). Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids. It has been shown to bind to single-stranded DNA (although not as strongly) and triple-stranded DNA. Because of its ability to bind to DNA, EtBr is an inhibitor of DNA polymerase.

Frameshift mutagen which intercalates double-stranded DNA and RNA.
Ethidium bromide has been used to prepare fluorescent stock solution for the evaluation of cell viability during the live/dead assay of adipose-derived stem cells (ADSCs) seeded on irradiated hydrogels.

Biochem/physiol Actions

Ethidium bromide intercalates double-stranded DNA and RNA and acts as a frameshift mutagen. It can also be used in conjunction with acridine orange to differentiate between viable, apoptotic and necrotic cells.

Reconstitution

For staining a gel after electrophoresis, dilute a sample of the stock solution to 0.5 μg/ml with water and incubate the gel for 15-30 min. Destaining is usually not needed but can be carried out in water for 15 min if decreased background is necessary. The DNA bands can then be detected on a UV light box (254 nm wavelength). Ethidium bromide can also be incorporated into the gel and running buffer at 0.5 μg/ml and visualized immediately after electrophoresis.

Other Notes

Intercalating agent and fluorescent label for DNA. Review; Destruction and decontamination of solutions

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Muta. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Lunn et al.
Analytical biochemistry, 162(2), 453-458 (1987-05-01)
Ethidium bromide in water, TBE buffer, Mops buffer, and cesium chloride solution may be completely degraded by reaction with sodium nitrite and hypophosphorous acid. Only non-mutagenic reaction mixtures were produced. Destruction was greater than 99.8% in all cases; the limit
W. Warring
Antibiotics III, 141-141 (1975)
Selective inhibition of the synthesis of mitochondria-associated RNA by ethidium bromide.
E Zylber et al.
Journal of molecular biology, 44(1), 195-204 (1969-08-28)
Complex formation between ethidium bromide and nucleic acids.
Waring MJ.
Journal of Microbiology, 13(1), 269-282 (1965)
A fluorescent complex between ethidium bromide and nucleic acids. Physical-chemical characterization.
J B LePecq et al.
Journal of molecular biology, 27(1), 87-106 (1967-07-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service