Skip to Content
Merck
All Photos(1)

Key Documents

45972

Supelco

2,4-Dichlorotoluene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3Cl2
CAS Number:
Molecular Weight:
161.03
Beilstein:
1931691
EC Number:
MDL number:
PubChem Substance ID:

grade

analytical standard

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.546 (lit.)

bp

200 °C (lit.)

density

1.246 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cc1ccc(Cl)cc1Cl

InChI

1S/C7H6Cl2/c1-5-2-3-6(8)4-7(5)9/h2-4H,1H3

InChI key

FUNUTBJJKQIVSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riquan Liao et al.
Environmental monitoring and assessment, 191(5), 309-309 (2019-04-28)
To enhance our understanding on environmental conditions of the Maowei Sea in Guangxi province, China, the concentration and distribution of 22 chlorobenzene compounds (CBs) in the surface sediment were determined by gas chromatography-mass spectrometry (GC-MS). The relationship of the sediment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service