Skip to Content
Merck
All Photos(1)

Key Documents

45240

Sigma-Aldrich

Eosin Y disodium salt

for microscopy (Fl., Hist.)

Synonym(s):

2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Acid Red 87, Bromo acid J. TS, XL, or XX, Bromofluorescein, Bronze Bromo ES, D&C;Red No. 22, Eosin yellowish

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H6Br4Na2O5
CAS Number:
Molecular Weight:
691.85
Colour Index Number:
45380
Beilstein:
3586809
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

for microscopy (Fl., Hist.)

form

powder

technique(s)

titration: suitable

solubility

H2O: may be turbid 0.1 g/10 mL, red to red-brown

density

1.018 g/cm3

εmax

≥1050 at 509-519 nm

fluorescence

λex 532 nm
λem 540 nm in H2O

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[O-]C(=O)c1ccccc1C2=C3C=C(Br)C(=O)C(Br)=C3Oc4c(Br)c([O-])c(Br)cc24

InChI

1S/C20H8Br4O5.2Na/c21-11-5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(22)17(26)15(24)19(10)29-18(9)14(23)16(11)25;;/h1-6,25H,(H,27,28);;/q;2*+1/p-2

InChI key

SEACYXSIPDVVMV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshihisa Yashiro et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(48), 17212-17217 (2014-11-19)
Males and females are in conflict over genetic transmission in the evolution of parthenogenesis, because it enhances female reproductive output but deprives the males' genetic contribution. For males, any trait that coerces females into sexual reproduction should increase their fitness.
Jie Du et al.
Fish & shellfish immunology, 34(1), 315-323 (2012-11-28)
Freshwater prawn Macrobrachium rosenbergii inoculated with 100 μl novel pathogen spiroplasma strain MR-1008 in logarithmic phase (10(8) spiroplasmas ml(-1)) were examined for alkaline phosphatase (AKP) activity, acid phosphatase (ACP) activity, superoxide dismutase (SOD) activity, catalase (CAT) activity, as well as
Patricia S Nielsen et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 26(3), 404-413 (2012-11-24)
Cellular proliferation is correlated with the progression of melanoma. Accordingly, the proliferation index of H&E-stained thin melanomas was recently included in the staging system of the American Joint Committee on Cancer. Yet, the immunohistochemical markers of proliferation phosphohistone H3 and
Sarah J Hatsell et al.
Science translational medicine, 7(303), 303ra137-303ra137 (2015-09-04)
Fibrodysplasia ossificans progressiva (FOP) is a rare genetic disorder characterized by episodically exuberant heterotopic ossification (HO), whereby skeletal muscle is abnormally converted into misplaced, but histologically normal bone. This HO leads to progressive immobility with catastrophic consequences, including death by
Han Shih et al.
ACS applied materials & interfaces, 5(5), 1673-1680 (2013-02-07)
Interfacial visible light-mediated thiol-ene photoclick reactions were developed for preparing step-growth hydrogels with multilayer structures. The effect of a noncleavage type photoinitiator eosin-Y on visible-light-mediated thiol-ene photopolymerization was first characterized using in situ photorheometry, gel fraction, and equilibrium swelling ratio.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service