Skip to Content
Merck
All Photos(1)

Key Documents

4466004

Sigma-Aldrich

CYP450 Extended Panel - Starter Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$4,080.00

$4,080.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 KIT
$4,080.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.24

$4,080.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

technique(s)

LC/MS: suitable

General description

The SCIEX iChemistry® CYP450 Peptide Standards Extended Panel Human Induction Starter Kit provides a complete package for the analysis of seven human CYP450 isoforms (1A2, 2B6, 2C9, 2C19, 2E1, 3A4, 3A5) using MRM analysis. The kit contains a detailed protocol, instrument data acquisition methods, processing methods, LC column, stable isotope-labeled synthetic peptide standards, trypsin, and all buffers and reagents for 100 assays.

Analysis Note

To view the Protocol for the CYP450 Protein Assay - Human Induction Kit Extended Panel, please visit this Protocol link.

A Technical Note on the use of the CYP450 Protein Assay - Human Induction Kit Extended Panel is available at this Tech Note link.

Legal Information

iChemistry is a registered trademark of AB Sciex Pte. Ltd.

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service