Skip to Content
Merck
All Photos(1)

Key Documents

44614

Sigma-Aldrich

Durcupan ACM

single component D

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$71.30

$71.30


Estimated to ship onMay 16, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$71.30

About This Item

EC Number:
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.25

$71.30


Estimated to ship onMay 16, 2025


Request a Bulk Order

Quality Level

form

liquid

contains

plasticizer

Looking for similar products? Visit Product Comparison Guide

Application

Embedding material for electron microscopy on the basis of Araldite®.

Legal Information

Araldite is a registered trademark of Huntsman Advanced Materials Inc.
Durcupan is a trademark of Sigma-Aldrich Chemie GmbH

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Repr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

367.7 °F - open cup

Flash Point(C)

186.5 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noemi Holderith et al.
Cell reports, 32(4), 107968-107968 (2020-07-30)
Elucidating the molecular mechanisms underlying the functional diversity of synapses requires a high-resolution, sensitive, diffusion-free, quantitative localization method that allows the determination of many proteins in functionally characterized individual synapses. Array tomography permits the quantitative analysis of single synapses but

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service