Skip to Content
Merck
All Photos(1)

Key Documents

43817

Sigma-Aldrich

DL-Dithiothreitol

BioUltra, ≥99.0% (RT)

Synonym(s):

threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25
Beilstein:
1719757
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioUltra

Assay

≥99.0% (RT)

form

solid

impurities

insoluble matter, passes filter test
≤0.3% 4,5-dihydroxy-1,2-dithiane

pH

4.0-6.5 (25 °C, 0.1 M in H2O)

mp

41-44 °C (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: 0.400
λ: 280 nm Amax: 0.100

storage temp.

2-8°C

SMILES string

O[C@H](CS)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m1/s1

InChI key

VHJLVAABSRFDPM-QWWZWVQMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

An excellent reagent for maintaining SH groups in reduced state; quantitatively reduces disulfides. DTT is effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N′-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. DTT is less pungent and is less toxic than 2-mercaptoethanol. Typically, a seven fold lower concentration of DTT (100 mM) is needed than is used for 2-mercaptoethanol (5% v/v, 700 mM).

Other Notes

Cleavage of disulfide bonds in proteins;[1] In the investigation of incremental and kinetic reduction in proteins;[2] Prevents the oxidation of cholecystokinin during its extraction from brain and duodenal tissues;[3] In protein sequence analysis.[4]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D W Sears et al.
Biochemistry, 16(9), 2031-2035 (1977-05-03)
The reduction by dithiothreitol (DTT) of the four interchain disulfides of a human IgGlkappa immunoglobulin has been studied by two methods: variation of the concentration of DTT relative to the protein concentration (incremental reduction); and variation of the time of
J R Jabusch et al.
Analytical biochemistry, 91(2), 532-542 (1978-12-01)
The thiazolinone and phenylthiohydantoin derivatives of most amino acids can be hydrolyzed with alkaline dithionite to generate the free amino acid. The acidification of this hydrolysate with 3 N HCl containing thiodiglycol leads in the case of glutamic acid, glutamine
A.Bacarese-Hamilton et al.
Annals of the New York Academy of Sciences, 448, 571-571 (1985)
Reductive cleavage of cystine disulfides with tributylphosphine.
U T Rüegg et al.
Methods in enzymology, 47, 111-116 (1977-01-01)
Xueqian Liu et al.
The Journal of biological chemistry, 288(10), 7230-7240 (2013-01-25)
In Pichia pastoris, the peroxisomal targeting signal 2 (PTS2)-dependent peroxisomal matrix protein import pathway requires the receptor, Pex7, and its co-receptor Pex20. A conserved lysine (Lys(19)) near the N terminus of Pex20 is required for its polyubiquitination and proteasomal degradation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service