437R-1
CD103 (EP206) Rabbit Monoclonal Primary Antibody
About This Item
biological source
rabbit
Quality Level
100
500
conjugate
unconjugated
antibody form
culture supernatant
antibody product type
primary antibodies
clone
EP206, monoclonal
description
(For In Vitro Diagnostic Use in Select Regions (See Chart))
form
buffered aqueous solution
species reactivity
human
packaging
bottle of 1.0 mL predilute (437R-17)
bottle of 7.0 mL predilute (437R-18)
vial of 0.1 mL concentrate (437R-14)
vial of 0.5 mL concentrate (437R-15)
vial of 1.0 mL concentrate (437R-16)
manufacturer/tradename
Cell Marque®
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100
isotype
IgG
shipped in
wet ice
storage temp.
2-8°C
Gene Information
human ... ITGAE(3682)
Related Categories
General description
Quality
![]() IVD | ![]() IVD | ![]() IVD | ![]() RUO |
Linkage
Physical form
Preparation Note
Other Notes
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service