Skip to Content
Merck
All Photos(1)

Key Documents

4352160

Sigma-Aldrich

iTRAQ® Reagents Methods Developments Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,790.00

$1,790.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 KIT
$1,790.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.24

$1,790.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

storage temp.

-10 to -25°C

General description

Contains iTRAQ® reagents 114, 117, test sample, buffers and reagents for up to three separate analyses. Each individual reagent can label up to 100 μg of protein. Included: Starter Kit - Hardware, Cation Exchange Buffer Pack and Cation Exchange Cartridge, Protocol, iTRAQ® Reagent Kit.

Analysis Note

To view the general iTRAQ Reagents protocol, please visit this Protocol link.

To view the Chemistry Quick Reference Card for the iTRAQ Reagents, please visit this Chemistry Quick Reference Card link.

Legal Information

iTraq is a registered trademark of AB Sciex Pte. Ltd.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service