Skip to Content
Merck
All Photos(1)

Key Documents

43407

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide hydrate

≥94% (HPLC), from yeast

Synonym(s):

β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7O14P2 · xH2O
CAS Number:
Molecular Weight:
663.43 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
39200202
PubChem Substance ID:

biological source

yeast

Assay

≥94% (HPLC)

impurities

≤10% water
≤4% solvent (methanol)

solubility

H2O: 0.1 g/mL, clear, faintly yellow
water: freely soluble

suitability

in accordance for determination of ADH

storage temp.

−20°C

SMILES string

NC1=NC=NC2=C1N=CN2.O[C@@H]3[C@@H](COP(O)(OP(OC[C@@H](O4)[C@@H](O)[C@@H](O)[C@H]4[N+]5=CC=CC(C(N)=O)=C5)([O-])=O)=O)OC[C@@H]3O

InChI

1S/C21H27N7O14P2/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H5-,22,23,24,25,33,34,35,36,37)/t10-,11-,13-,14-,15-,16-,20-,21-/m1/s1

InChI key

BAWFJGJZGIEFAR-NNYOXOHSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

β-Nicotinamide adenine dinucleotide hydrate was used as a test compound for studying the structure and electrochemical properties of NADH electrocatalysts.

Biochem/physiol Actions

β-Nicotinamide adenine dinucleotide is a cofactor for alcohol dehydrogenase that serves as an inhibitory neurotransmitter in visceral smooth muscle.
Electron acceptor

Packaging

Packaged by solid weight.

Other Notes

This is the common form of NAD.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and Electrochemical Properties of Electrocatalysts for NADH Oxidation.
Rosalba A. Rincon
Electroanalysis, 22, 799-806 (2010)
Violeta N Mutafova-Yambolieva et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(41), 16359-16364 (2007-10-05)
Peripheral inhibitory nerves are physiological regulators of the contractile behavior of visceral smooth muscles. One of the transmitters responsible for inhibitory neurotransmission has been reputed to be a purine, possibly ATP. However, the exact identity of this substance has never
Christian Dölle et al.
The FEBS journal, 280(15), 3530-3541 (2013-04-27)
Mitochondrial metabolism is intimately connected to the universal coenzyme NAD. In addition to its role in redox reactions of energy transduction, NAD serves as substrate in regulatory reactions that lead to its degradation. Importantly, all types of the known NAD-consuming
Jeerus Sucharitakul et al.
The Journal of biological chemistry, 288(49), 35210-35221 (2013-10-17)
3-Hydroxybenzoate 6-hydroxylase (3HB6H) from Rhodococcus jostii RHA1 is an NADH-specific flavoprotein monooxygenase that catalyzes the para-hydroxylation of 3-hydroxybenzoate (3HB) to form 2,5-dihydroxybenzoate (2,5-DHB). Based on results from stopped-flow spectrophotometry, the reduced enzyme-3HB complex reacts with oxygen to form a C4a-peroxy
Ana P Gomes et al.
Cell, 155(7), 1624-1638 (2013-12-24)
Ever since eukaryotes subsumed the bacterial ancestor of mitochondria, the nuclear and mitochondrial genomes have had to closely coordinate their activities, as each encode different subunits of the oxidative phosphorylation (OXPHOS) system. Mitochondrial dysfunction is a hallmark of aging, but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service