Skip to Content
Merck
All Photos(1)

Key Documents

42137

Sigma-Aldrich

Lactosylsphingosine

≥98.0% (TLC)

Synonym(s):

(2S,3R,4E)-2-Amino-3-hydroxy-4-octadecen-1-yl 4-O-β-D-galactopyranosyl-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H57NO12
CAS Number:
Molecular Weight:
623.77
Beilstein:
5680460
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.85

Assay

≥98.0% (TLC)

form

powder

lipid type

sphingolipids

storage temp.

−20°C

SMILES string

OC1[C@@H](O)[C@H](OC[C@@H](N)[C@@H](O)/C=C/CCCCCCCCCCCCC)OC(CO)[C@H]1O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)[C@@H](CO)O2

InChI

1S/C30H57NO12/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-20(34)19(31)18-40-29-27(39)25(37)28(22(17-33)42-29)43-30-26(38)24(36)23(35)21(16-32)41-30/h14-15,19-30,32-39H,2-13,16-18,31H2,1H3/b15-14+/t19-,20+,21?,22?,23+,24-,25-,26?,27-,28?,29-,30+/m1/s1

InChI key

MQKSCOKUMZMISB-VOCCQAPRSA-N

Biochem/physiol Actions

Lactosylsphingosine is relevant for cancer, as lactosyl-reative antibodies occur in sera of cancer patients.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xavier Buton et al.
Biochemistry, 41(43), 13106-13115 (2002-10-23)
The transbilayer movement of glycosphingolipids has been characterized in Golgi, ER, plasma, and model membranes using spin-labeled and fluorescent analogues of the monohexosylsphingolipids glucosylceramide and galactosylceramide and of the dihexosylsphingolipid lactosylceramide. In large unilamellar lipid vesicles, monohexosylsphingolipids underwent a slow
Takehiro Nagatsuka et al.
ACS applied materials & interfaces, 5(10), 4173-4180 (2013-05-15)
We have detected biological toxins using localized surface plasmon resonance (LSPR) and synthetic glycosyl ceramides (β-lactoside, globosyl trisaccharide (Gb3), or GM1 pentasaccharide) attached to gold (Au) nanoparticles. The particle diameters ranged from 5-100 nm. The detection sensitivity for three toxins
Kevin Mills et al.
Rapid communications in mass spectrometry : RCM, 19(12), 1739-1748 (2005-05-24)
Novel internal standards have been synthesised for the quantitative determination by tandem mass spectrometry (MS/MS) of the sphingolipids that accumulate in lysosomal storage diseases. The [d4]C16- and [d47]C24-isoforms of galactosylceramide (CMH), lactosylceramide (CDH), globotriaosylceramide (CTH), cerebroside sulphate, sphingomyelin and G(M1)-
Sumita Mishra et al.
Glycobiology, 24(6), 518-531 (2014-03-25)
Hypertrophy is central to several heart diseases; however, not much is known about the role of glycosphingolipids (GSLs) in this phenotype. Since GSLs have been accorded several physiological functions, we sought to determine whether these compounds affect cardiac hypertrophy. By
Margarita L Malakhova et al.
The Journal of biological chemistry, 280(28), 26312-26320 (2005-05-20)
Mammalian glycolipid transfer proteins (GLTPs) facilitate the selective transfer of glycolipids between lipid vesicles in vitro. Recent structural determinations of the apo- and glycolipid-liganded forms of human GLTP have provided the first insights into the molecular architecture of the protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service