Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC (p)
Citations:
2
Pricing and availability is not currently available.
biological source
rabbit
conjugate
unconjugated
antibody form
Ig fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
description
For In Vitro Diagnostic Use in Select Regions (See Chart)
form
buffered aqueous solution
species reactivity
human
packaging
vial of 0.1 mL concentrate (413A-14), vial of 0.5 mL concentrate (413A-15), bottle of 1.0 mL predilute (413A-17), vial of 1.0 mL concentrate (413A-16), bottle of 7.0 mL predilute (413A-18)
manufacturer/tradename
Cell Marque®
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100
control
embryonal carcinoma, yolk sac tumor
shipped in
wet ice
storage temp.
2-8°C
visualization
membranous
Gene Information
human ... SLC2A3(6515)
General description
Glucose transporter membrane 3 (GLUT3) is known as a solute carrier family 2 (facilitated glucose transporter) member 3 and represents a membrane bound glucose transporter. In one study scientists, using immunoprecipitation and western blot technologies, detected GLUT3 expression in spermatozoa of testis and brain but not in other types of tissue.[1] In-house research via immunohistochemistry confirmed GLUT3 expression in testis/spermatozoa, but no GLUT3 was detected in brain. In a study conducted by Howitt BE et. al. anti-GLUT3 demonstrated membranous staining of seminoma cells in 44 cases (65%) of total 67 specimens.[2] In addition, anti-GLUT3 labeled embryonal carcinoma (20/20) and yolk sac tumor (8/8) with 100% sensitivity.[2]GLUT3 is not expressed in non-germ cell tumors such as leydig cell tumor and adenomatoid tumor.[2]Spermatocytic seminoma, choriocarcinoma, and immature teratoma are also negative for GLUT3.[2] Therefore, anti-GLUT3 is a very useful IHC marker to include in a panel for identification of germ cell tumors.
Physical form
Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide
Preparation Note
Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.
Analysis Note
![]() IVD | ![]() IVD | ![]() IVD | ![]() RUO |
Other Notes
For Technical Service please contact: 800-665-7284 or email: [email protected]
GLUT3 Positive Control Slides, Product No. 413S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).
Legal Information
Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 413A-17 | 0841683141723 |
| 413A-18 | 0841683141730 |
| 413A-16 | 0841683141716 |
| 413A-15 | 0841683141709 |
| 413A-14 | 0841683141693 |



