Skip to Content
Merck
All Photos(1)

Key Documents

411-500

Sigma-Aldrich

Chondrocyte Growth Medium (500 ml)

Synonym(s):

Primary Cell Culture Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
Pricing and availability is not currently available.

form

liquid

Quality Level

shelf life

6 mo.

manufacturer/tradename

Cell Applications, Inc

shipped in

wet ice

storage temp.

2-8°C

General description

All-in-one ready-to-use media

Application

Chondrocyte Growth Medium (500 ml) has been used in culturing of human:
  • growth plate chondrocytes[1]
  • osteoarthritic chondrocytes[2]
  • articular chondrocytes[3]

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Xue et al.
Molecular medicine reports, 8(6), 1755-1760 (2013-10-16)
Tumor necrosis factor (TNF)-α and a disintegrin and metalloproteinase with thrombospondin motifs 4 (ADAMTS-4) are important in osteoarthritis (OA) cartilage degradation. In the present study, we explored the interaction between the two proteins by examining the effect of TNF-α on
Bing Wang et al.
Bone, 146, 115875-115875 (2021-02-12)
In our previous study, the mutation c.2645A > C (p. E882A) was found in the A-Kinase Anchoring Protein 2 (AKAP2) gene, which plays an important role in regulating the development of the skeletal system; however, the specific effect of AKAP2 on chondrocyte
Rachel L Price et al.
Biomaterials, 24(11), 1877-1887 (2003-03-05)
Bone cell adhesion on novel carbon nanofibers and polycarbonate urethane/carbon nanofiber (PCU/CNF) composites is investigated in the present in vitro study. Carbon nanofibers have exceptional theoretical mechanical properties (such as high strength to weight ratios) that, along with possessing nanoscale

Protocols

Cryovial handling precautions prevent injuries from liquid nitrogen bursts.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service