Skip to Content
Merck
All Photos(1)

Key Documents

394R-1

Sigma-Aldrich

Stathmin (SP49) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP49, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (394R-14)
vial of 0.5 mL concentrate (394R-15)
bottle of 1.0 mL predilute (394R-17)
vial of 1.0 mL concentrate (394R-16)
bottle of 7.0 mL predilute (394R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG

control

cervical intraepithelial neoplasia – high grade, tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... STMN1(3925)

General description

The distinction between high grade cervical intraepithelial neoplasia (CIN2/3) from low grade cervical intraepithelial neoplasia (CIN1) is clinically significant with treatment recommendations linked specificallyto risk of cancer or CIN3 outcome.[1] CIN1 will progress into invasive carcinoma in <1% of cases and is typically managed with Papanicolaou smear follow-up, whereas CIN2/3 has a 5% to 20% risk of progression1 and is usually treated with an excisional procedure (loop electrosurgical excision procedure or cone biopsy).1 Stathmin, also referred to as Stathmin-1 and oncoprotein18, is a ubiquitous microtubule-destabilizing protein shown to be important during mitosis and has been implicated as a regulator of cell motility and migration.[2]

Recent studies[3] show anti-stathmin is positive in 24/82 (29%) CINs with differential expression based on the grade of the lesion as 5/56 (9%) CIN1, 5/11 (45%) CIN2, and 14/15 (93%) CIN3; whereas, anti-p16 staining of the same cases was immuno-reactive in 66/83 (80%) CINs, including 40/56 (71%) CIN1, 11/11 (100%) CIN2, and 15/16 (94%) CIN3.[3] Anti-stathmin shows similar sensitivity for CIN3 to anti-p16 (93% vs 94%) although it drops off for CIN2 (73% vs 96%). The specificity of anti-stathmin for both CIN2/3 (94%) and CIN3 (89%) is higher than that of anti-p16 (44% and 39%, respectively). Anti-Stathmin stains basal layer of normal benign ectocervix. A well-oriented fragment of cervix tissue would increase the accuracy of diagnosis. In conclusion, based on recent studies, anti-stathmin has a higher specificity relative to anti-p16; therefore, anti-stathmin has major potential as a diagnostic marker in CIN classification over anti-p16.[3]

Quality


IVD

IVD

IVD

RUO

Linkage

Stathmin Positive Control Slides, Product No. 394S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Syrjänen
European journal of obstetrics, gynecology, and reproductive biology, 65(1), 45-53 (1996-03-01)
Because of the fact that any meaningful classification should bear a close relationship to the biological behavior of the lesions, the usefulness of all new classifications of cervical precancer lesions can only be established by well controlled prospective follow-up studies.
Bellal Joseph et al.
American journal of surgery, 207(1), 89-94 (2013-10-15)
Patients with fatal gunshot wounds (GSWs) to the head often have poor outcomes but are ideal candidates for organ donation. The purpose of this study was to evaluate the effects of aggressive management on organ donation in patient with fatal
Barbara Belletti et al.
Expert opinion on therapeutic targets, 15(11), 1249-1266 (2011-10-08)
Stathmin is a microtubule-destabilizing phosphoprotein, firstly identified as the downstream target of many signal transduction pathways. Several studies then indicated that stathmin is overexpressed in many types of human malignancies, thus deserving the name of Oncoprotein 18 (Op18). At molecular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service