Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.47
UNSPSC Code:
12352106
Pricing and availability is not currently available.
form
powder
Quality Level
shelf life
Expiry date on the label.
IVD
for in vitro diagnostic use
pH
7.0 (20 °C)
application(s)
hematology
histology
shipped in
wet ice
storage temp.
2-8°C
Application
Kit component in the Sigma Leukocyte Peroxidase (Myeloperoxidase) Procedure, 390A.
Other Notes
Contains p-Phenylenediamine diHCL (1 part) and catechol (2 parts). Commonly referred to as Hanker Yates Reagent.
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yuta Enami et al.
Gastroenterology, 136(7), 2356-2364 (2009-03-24)
Hepatic inflammation occurs immediately after cells are transplanted to the liver, but the mechanisms that underlie this process are not fully defined. We examined cyclooxygenase pathways that mediate hepatic inflammation through synthesis of prostaglandins, prostacyclins, thromboxanes, and other prostanoids following
Lilianne Barbar et al.
Neuron, 107(3), 436-453 (2020-06-03)
New methods for investigating human astrocytes are urgently needed, given their critical role in the central nervous system. Here we show that CD49f is a novel marker for human astrocytes, expressed in fetal and adult brains from healthy and diseased
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 3901-10VL | 04061838202895 |



