Skip to Content
Merck
All Photos(1)

Documents

389M-1

Sigma-Aldrich

Glutamine Synthetase (GS-6) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352116
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

GS-6, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (389M-14)
vial of 0.5 mL concentrate (389M-15)
bottle of 1.0 mL predilute (389M-17)
vial of 1.0 mL concentrate (389M-16)
bottle of 7.0 mL predilute (389M-18)

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG2a

control

hepatocellular carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... GLUL(2752)

General description

Glutamine synthetase (GS) catalyzes the synthesis of glutamine from glutamate and ammonia in the mammalian liver. In normal liver, GS expression is seen in centrilobular (zone 3) hepatocytes, but not in mid-zone (zone 2) or periportal (zone 3) hepatocytes. Glutamine synthetase immunohistochemistry is also positive in neoplasms of hepatocyte origin, such as hepatocellular carcinoma (HCC) and hepatocellular adenoma, making it useful for identifying these tumors.

Quality


IVD

IVD

IVD

RUO

Linkage

Glutamine Synthetase Positive Control Slides, Product No. 389S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luca Di Tommaso et al.
Hepatology (Baltimore, Md.), 45(3), 725-734 (2007-02-28)
Hepatocellular nodules in cirrhosis include regenerative (large regenerative, LRN) and dysplastic (low and high grade, LGDN and HGDN) nodules, early and grade 1 HCC (eHCC-G1), and overt HCC. The differential diagnosis may be particularly difficult when lesions such as HGDN
Jiakun Chen et al.
Neuron, 112(1), 93-112 (2023-12-15)
Astrocytes play crucial roles in regulating neural circuit function by forming a dense network of synapse-associated membrane specializations, but signaling pathways regulating astrocyte morphogenesis remain poorly defined. Here, we show the Drosophila lipid-binding G protein-coupled receptor (GPCR) Tre1 is required

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service