Skip to Content
Merck
All Photos(2)

Key Documents

36792

Supelco

Aclonifen

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-6-nitro-3-phenoxyaniline

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$80.40

$80.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$80.40

About This Item

Empirical Formula (Hill Notation):
C12H9ClN2O3
CAS Number:
Molecular Weight:
264.66
Beilstein:
8321574
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$80.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1c(Cl)c(Oc2ccccc2)ccc1[N+]([O-])=O

InChI

1S/C12H9ClN2O3/c13-11-10(18-8-4-2-1-3-5-8)7-6-9(12(11)14)15(16)17/h1-7H,14H2

InChI key

DDBMQDADIHOWIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aclonifen is a diphenyl-ether herbicide, which can be used for weed control management, in specific for sunflowers.[1] Its mode of action involves, the inhibition of carotenoid synthesis and suppression of protoporphyrinogen oxidase enzyme involved in the chlorophyll synthesis pathway.[1]

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Dhouib et al.
Central European journal of public health, 4 Suppl, 51-52 (1996-01-01)
The herbicides Alachlor and Alclonifen are recently used in France for pre-emergent weed control in corn, essentially in association. For this reason, an improved method has been developed by gas chromatography for simultaneously characterising and quantifying the two herbicides. The
James A Garratt et al.
Pest management science, 59(1), 3-20 (2003-02-01)
Effective prediction of pesticide fate using mathematical models requires good process descriptions in the models and good choice of parameter values by the user. This paper examines the ability of seven pesticide leaching models (LEACHP, MACRO, PELMO, PESTLA, PLM, PRZM
The herbicide aclonifen: The complex theoretical bases of sunflower tolerance
Kilinc O, et al.
Pesticide Biochemistry and Physiology, 100, 193-198 (2011)
Praveen Kumar Gopi et al.
Ecotoxicology and environmental safety, 207, 111285-111285 (2020-09-16)
The present work scrutinized the voltammetric analysis of hazardous herbicide aclonifen (ACF) in actual soil and river water samples utilizing the electrochemical method. The electrochemical sensing device was fabricated for the determination of ACF using gadolinium niobate (GdNbO4) nanoparticles modified
L Jacquin et al.
Ecotoxicology and environmental safety, 172, 255-264 (2019-02-04)
In human-altered rivers, fish are often conjointly exposed to an increase in water temperature due to global warming and to a contamination by organic pollutants such as pesticides, but their combined effects are still elusive. Thermal and chemical stressors could

Questions

1–2 of 2 Questions  
  1. What is the molecular formula for the smaller molecules of Product 36792?

    1 answer
    1. The molecular formula for this product is C12H9ClN2O3.

      Helpful?

  2. What is the molecular formula for smaller molecules?

    1 answer
    1. The molecular formula for this product is C12H9ClN2O3.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service