Skip to Content
Merck
All Photos(1)

Key Documents

34022

Trizma® buffer solutions 0.1 M

pH 7.2 (37 °C), pH 7.5 (25 °C), pH 8.1 (5 °C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51000000

IVD

for in vitro diagnostic use

pH

7.2 (37 °C)
7.5 (25 °C)
8.1 (5 °C)

storage temp.

2-8°C

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service