Skip to Content
Merck

314331

Toluene-(methyl-14C)

≥98% (GC, HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H514CH3
CAS Number:
Molecular Weight:
92.14
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (GC, HPLC)

extent of labeling

1-15 mCi per mmol

storage temp.

2-8°C

SMILES string

[14CH3]c1ccccc1

Packaging

Packaged in break-seal ampule




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library