Skip to Content
Merck
All Photos(1)

Key Documents

31191

Sigma-Aldrich

D.E.R. 736

Synonym(s):

Epoxy-Resin

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$192.00

$192.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
$192.00

About This Item

UNSPSC Code:
12352106
NACRES:
NA.25

$192.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

density

1.14 g/mL at 20 °C

General description

a shorter chain aliphatic diepoxide than D.E.R. 732; gives slightly less flexibility, impact and elongation with some improvement in H.D.T. and chemical resistance; with Lemo connector

Legal Information

D.E.R. is a trademark of The Dow Chemical Company or an affiliated company of Dow

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A new embedding method employing D.E.R. 736 and epon 812.
H Kushida
Journal of electron microscopy, 16(3), 278-280 (1967-01-01)
Jun Nie et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(3), 1901891-1901891 (2020-02-12)
The recent integration of light-sheet microscopy and tissue-clearing has facilitated an important alternative to conventional histological imaging approaches. However, the in toto cellular mapping of neural circuits throughout an intact mouse brain remains highly challenging, requiring complicated mechanical stitching, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service