Skip to Content
Merck
All Photos(1)

Documents

272M-1

Sigma-Aldrich

INI-1 (MRQ-27) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-27, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (272M-14)
vial of 0.5 mL concentrate (272M-15)
bottle of 1.0 mL predilute (272M-17)
vial of 1.0 mL concentrate (272M-16)
bottle of 7.0 mL predilute (272M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

control

brain, endothelial cells

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... SMARCB1(6598)

General description

INI-1, also known as SMARCB1, is one of the core subunit proteins of the ATP-dependent SWI/SNF chromatin remodeling complex. It is involved in chromatin remodeling and transcriptional regulation and functions as a tumor suppressor. Nuclear INI-1 expression is seen in virtually all normal human tissues and most malignancies but the INI-1 gene is often mutated or deleted in malignant rhabdoid tumors (MRT), including those arising in the central nervous system, leading to a lack of INI-1 expression.

Quality


IVD

IVD

IVD

RUO

Linkage

INI-1 Positive Control Slides, Product No. 272S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Fowler et al.
Fetal and pediatric pathology, 25(3), 159-168 (2006-10-25)
Primary extrarenal rhabdoid tumors (RT) are now recognized as a specific entity in pediatric oncological pathology practice. We present an unusual case of a small cell myxoid variant of a thoracic RT in an infant and highlight the importance of
Christine Haberler et al.
The American journal of surgical pathology, 30(11), 1462-1468 (2006-10-26)
Immunohistochemical lack of nuclear INI1 protein expression has been recently described as characteristic finding in atypical teratoid/rhabdoid tumors (AT/RTs), and has been suggested as useful marker to distinguish AT/RTs from other malignant pediatric central nervous system (CNS) tumors. In this
F Bourdeaut et al.
The Journal of pathology, 211(3), 323-330 (2006-12-08)
Rhabdoid tumours (RTs) are rare but highly aggressive tumours of childhood. Their rarity and their miscellaneous locations make the diagnosis particularly challenging for pathologists. Central nervous system and peripheral RTs have been associated with biallelic inactivation of the hSNF5/INI1/SMARCB1 (hSNF5/INI1)
Linlin Hao et al.
Oncology letters, 26(4), 428-428 (2023-09-04)
Multiple primary malignant neoplasms (MPMN) are defined as two or more primary malignancies diagnosed in an individual. There is no association between these cancers, which can be classified into synchronous and heterochronous cancers depending on the time of diagnosis. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service