Skip to Content
Merck
All Photos(1)

Key Documents

23120

Sigma-Aldrich

α-Chloralose

≥98.0% (chloralose, AT)

Synonym(s):

1,2-O-(2,2,2-Trichloroethylidene)-α-D-glucofuranose, Anhydro-D-glucochloral, Chloralose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11Cl3O6
CAS Number:
Molecular Weight:
309.53
Beilstein:
85418
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (chloralose, AT)

form

powder

optical activity

[α]20/D +17±2°, 5 hr, c = 2% in ethanol

impurities

15% Beta-chloralose

mp

178-184 °C

solubility

water: soluble 1 gm in 225 ml at 15 °C
chloroform: slightly soluble
diethyl ether: soluble
glacial acetic acid: soluble
petroleum ether: insoluble

SMILES string

OC[C@@H](O)[C@H]1O[C@@H]2O[C@@H](O[C@@H]2[C@H]1O)C(Cl)(Cl)Cl

InChI

1S/C8H11Cl3O6/c9-8(10,11)7-16-5-3(14)4(2(13)1-12)15-6(5)17-7/h2-7,12-14H,1H2/t2-,3+,4-,5-,6-,7-/m1/s1

InChI key

OJYGBLRPYBAHRT-IPQSZEQASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

α-Chloralose stimulates the gama-aminobutyric acid (GABA)-induced chloride conductance of gama-aminobutyric acidA (GABAA) receptors expressed in Xenopus laevis oocytes.[1]

Components

mixture of isomers of ~85% α-chloralose; ~15% β-chloralose

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R W Gear et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 19(16), 7175-7181 (1999-08-06)
We tested the hypothesis that noxious stimuli induce pain modulation by activation of supraspinal structures. We found that intense noxious stimuli (i.e., subdermal injection of capsaicin or paw immersion in hot water) induced profound attenuation of the jaw-opening reflex in
K M Garrett et al.
The Journal of pharmacology and experimental therapeutics, 285(2), 680-686 (1998-05-15)
alpha-Chloralose is widely used as an anesthetic in the laboratory due to its minimal effects on autonomic and cardiovascular systems, yet little is known about its mechanism of action. We examined the effects of alpha-chloralose on gamma-aminobutyric acid type A
Hongxia Lei et al.
Journal of neuroscience research, 88(2), 413-419 (2009-09-12)
Barbiturates are regularly used as an anesthetic for animal experimentation and clinical procedures and are frequently provided with solubilizing compounds, such as ethanol and propylene glycol, which have been reported to affect brain function and, in the case of (1)H
Renata F Leoni et al.
NeuroImage, 58(1), 75-81 (2011-06-29)
Hypertension afflicts 25% of the general population and over 50% of the elderly. In the present work, arterial spin labeling MRI was used to non-invasively quantify regional cerebral blood flow (CBF), cerebrovascular resistance and CO(2) reactivity in spontaneously hypertensive rats
Mingming Zhu et al.
Journal of comparative physiology. B, Biochemical, systemic, and environmental physiology, 179(6), 701-710 (2009-03-12)
Direct measurements of deep-brain and body-core temperature were performed on rats to determine the influence of cerebral blood flow (CBF) on brain temperature regulation under static and dynamic conditions. Static changes of CBF were achieved using different anesthetics (chloral hydrate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service