Skip to Content
Merck
All Photos(1)

Documents

22680

Sigma-Aldrich

Quinoline Yellow

for microscopy (Hist.), mixture of mono- and disulfonic acid sodium salt

Synonym(s):

Acid Yellow 3

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Colour Index Number:
47005
UNSPSC Code:
12171500
NACRES:
NA.47

grade

for microscopy (Hist.)

form

powder

solubility

H2O: 0.1 g/10 mL

εmax

≥350 at 283-293 nm in water
≥450 at 407-417 nm in water

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

InChI

1S/C18H11NO8S2.2Na/c20-17-11-3-1-2-4-12(11)18(21)15(17)13-6-5-9-7-10(28(22,23)24)8-14(16(9)19-13)29(25,26)27;;/h1-8,15H,(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2

InChI key

FZUOVNMHEAPVBW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian Weisz et al.
Journal of mass spectrometry : JMS, 37(10), 1025-1033 (2002-10-11)
Several positional isomers of 2-(2-quinolinyl)-1H-indene-1,3(2H)-dione mono- and disulfonic acids prepared as reference materials for development of analytical methods involved in FDA certification of D&C Yellow No. 10 (Quinoline Yellow) were found consistently to show [MH + 14](+) ions when their
Hisao Oka et al.
Journal of chromatography. A, 989(2), 249-255 (2003-03-26)
Quinoline yellow (Color Index No. 47005) consists of multiple components that show a large difference in their partition coefficients (K), ranging from 0.03 to 3.3 in the solvent system tert.-butyl methyl ether (MTBE)-1-butanol-acetonitrile-aqueous 0.1 M trifluoroacetic acid (TFA). Consequently, it
M Florian et al.
Food additives and contaminants, 19(9), 803-809 (2002-10-25)
A study of the voltammetric behaviour of the food colours brilliant blue FCF (C.I. 42090), erythrosine (C.I. 45430) and quinoline yellow (C.I. 47005) in the pH range 2-10 have been carried out by cathodic#10; stripping voltammetry. At pH 4.5 (acetate
Iatrogenic xanthoderma.
Nancy V Bruckner
Journal of the American Academy of Dermatology, 58(5), 895-896 (2008-04-22)
Karen Lau et al.
Toxicological sciences : an official journal of the Society of Toxicology, 90(1), 178-187 (2005-12-15)
Exposure to non-nutritional food additives during the critical development window has been implicated in the induction and severity of behavioral disorders such as attention deficit hyperactivity disorder (ADHD). Although the use of single food additives at their regulated concentrations is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service