Skip to Content
Merck
All Photos(1)

Key Documents

19459

Sigma-Aldrich

Protein G recombinant

from Escherichia coli, ≥90%

Synonym(s):

IgG F(c)-Receptor

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$319.00
5 MG
$1,260.00

$319.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$319.00
5 MG
$1,260.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$319.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

biological source

Escherichia coli

mol wt

Mr ~31800

concentration

≥90%

storage temp.

−20°C

General description

Protein G is a small globular cell surface protein produced by Streptococcus sp. It is composed of two or three nearly identical domains of 55 amino acids each. It binds to Fc fragment of IgG from many species with high affinity. It does not recognize IgM, IgA, IgE or serum albumin.

Physical form

lyophilized powder in PBS, pH 7.4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imke A K Burmester et al.
British journal of pharmacology, 177(22), 5114-5130 (2020-08-21)
Pemphigus is caused by autoantibodies against desmoglein (Dsg) 1, Dsg3, and/or non-Dsg antigens. Pemphigus vulgaris (PV) is the most common manifestation of pemphigus, with painful erosions on mucous membranes. In most cases, blistering also occurs on the skin, leading to
Robert Carlisle et al.
Pharmaceutical research, 30(2), 352-361 (2012-09-21)
To improve the delivery of liposomes to tumors using P-selectin glycoprotein ligand 1 (PSGL1) mediated binding to selectin molecules, which are upregulated on tumorassociated endothelium. PSGL1 was orientated and presented on the surface of liposomes to achieve optimal selectin binding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service