Skip to Content
Merck
All Photos(1)

Key Documents

19337

Sigma-Aldrich

Lectin from Ulex europaeus-Atto 488 conjugate

Synonym(s):

Ulex europaeus agglutinin-Atto 488 conjugate

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$2,020.00

$2,020.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 μG
$2,020.00

About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

$2,020.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

carbohydrate specificity α-L-Fucose

form

powder

fluorescence

λex 501 nm; λem 523 nm in PBS

storage temp.

−20°C

Application

Atto labels are designed for high sensitivity applications including fluorescence microscopy, flow cytometry, fluorescence in situ hybridization (FISH), receptor binding assays, or enzyme assays. Ulex europaeus agglutinin-Atto 488 conjugate, a green-emitting donor, may be used as FRET partner and as a marker of lectin binding.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina Scheller et al.
Tissue engineering. Part C, Methods, 19(1), 57-67 (2012-07-18)
A general problem in tissue engineering is the poor and insufficient blood supply to guarantee tissue cell survival as well as physiological tissue function. To address this limitation, we have developed an in vitro vascularization model in which a decellularized
Oliver J Wilson et al.
Histochemistry and cell biology, 138(4), 617-626 (2012-07-04)
Within animal skeletal muscle, focal adhesion kinase (FAK) has been associated with load-dependent molecular and metabolic adaptation including the regulation of insulin sensitivity. This study aimed to generate the first visual images of the localisation of FAK within human skeletal
María A Duque-Correa et al.
International journal for parasitology, 50(9), 707-718 (2020-07-14)
The caecum, an intestinal appendage in the junction of the small and large intestines, displays a unique epithelium that serves as an exclusive niche for a range of pathogens including whipworms (Trichuris spp.). While protocols to grow organoids from small
Yoshihiro Nishijima et al.
Genes to cells : devoted to molecular & cellular mechanisms, 17(10), 826-836 (2012-09-11)
Cell surface glycans change during the process of malignant transformation. To characterize and distinguish endometrial cancer and endometrium, we performed glycan profiling using an emerging modern technology, lectin microarray analysis. The three cell lines, two from endometrial cancers [well-differentiated type
Lei Chen et al.
Nature genetics, 51(5), 777-785 (2019-04-17)
BMP/SMAD signaling is a crucial regulator of intestinal differentiation1-4. However, the molecular underpinnings of the BMP pathway in this context are unknown. Here, we characterize the mechanism by which BMP/SMAD signaling drives enterocyte differentiation. We establish that the transcription factor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service