Skip to Content
Merck
All Photos(1)

Key Documents

18775

Sigma-Aldrich

AZCL-Hydroxyethylcellulose

substrate for celllulase

Synonym(s):

Azurine-cross-linked hydroxyethylcellulose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

powder

storage temp.

2-8°C

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service