Skip to Content
Merck
All Photos(1)

Key Documents

17195

Sigma-Aldrich

Fura 2 pentapotassium salt

for fluorescence, ≥90% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$562.00

$562.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$562.00

About This Item

Empirical Formula (Hill Notation):
C29H22K5N3O14
CAS Number:
Molecular Weight:
831.99
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

$562.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

grade

for fluorescence

Quality Level

Assay

≥90% (HPLC)

form

solid

solubility

H2O: soluble

fluorescence

λex 355 nm; λem 495 nm in 0.1 M Tris pH 8.0, Ca2+

storage temp.

−20°C

SMILES string

[K+].[K+].[K+].[K+].[K+].Cc1ccc(N(CC([O-])=O)CC([O-])=O)c(OCCOc2cc3cc(oc3cc2N(CC([O-])=O)CC([O-])=O)-c4ncc(o4)C([O-])=O)c1

InChI

1S/C29H27N3O14.5K/c1-15-2-3-17(31(11-24(33)34)12-25(35)36)20(6-15)43-4-5-44-21-7-16-8-22(28-30-10-23(46-28)29(41)42)45-19(16)9-18(21)32(13-26(37)38)14-27(39)40;;;;;/h2-3,6-10H,4-5,11-14H2,1H3,(H,33,34)(H,35,36)(H,37,38)(H,39,40)(H,41,42);;;;;/q;5*+1/p-5

InChI key

HQKWBBCTCLUGSX-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Other Notes

A very sensitive fluorescent calcium chelator that exhibits a spectral shift in excitation maxima on binding calcium[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of Ca2+ release in skinned fibers from skeletal muscle.
M Endo et al.
Methods in enzymology, 157, 12-26 (1988-01-01)
G Grynkiewicz et al.
The Journal of biological chemistry, 260(6), 3440-3450 (1985-03-25)
A new family of highly fluorescent indicators has been synthesized for biochemical studies of the physiological role of cytosolic free Ca2+. The compounds combine an 8-coordinate tetracarboxylate chelating site with stilbene chromophores. Incorporation of the ethylenic linkage of the stilbene

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service