Skip to Content
Merck
All Photos(1)

Key Documents

15000

Sigma-Aldrich

Bismarck Brown R

for microscopy (Bact., Hist.)

Synonym(s):

Basic Brown 4, Vesuvine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24N8 · 2HCl
CAS Number:
Molecular Weight:
461.39
Colour Index Number:
21010
Beilstein:
1829673
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy (Bact., Hist.)

form

powder or crystals

mp

222 °C (dec.) (lit.)

solubility

methanol: 0.01 g/10 mL, red

εmax

≥250 at 463-473 nm in 50% ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl[H].Cl[H].Cc1cc(\N=N\c2ccc(C)c(c2)\N=N\c3cc(C)c(N)cc3N)c(N)cc1N

InChI

1S/C21H24N8.2ClH/c1-11-4-5-14(26-28-20-6-12(2)15(22)9-17(20)24)8-19(11)27-29-21-7-13(3)16(23)10-18(21)25;;/h4-10H,22-25H2,1-3H3;2*1H/b28-26+,29-27+;;

InChI key

WLKAMFOFXYCYDK-CYXDXXRZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bismark Brown R (BB), also called as basic brown 4, belongs to the class of diazo dyes. It has the absorption maximum at 468nm.[1]

Application

Bismarck Brown R has been used as an adsorbate and is used in dye decoloration experiment.[2][1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Veronica Francardo et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 16(2), 465-479 (2019-02-14)
Pridopidine is a small molecule in clinical development for the treatment of Huntington's disease. It was recently found to have high binding affinity to the sigma-1 receptor, a chaperone protein involved in cellular defense mechanisms and neuroplasticity. Here, we have
John O Unuofin et al.
Journal of environmental management, 231, 222-231 (2018-10-21)
Agroindustrial residual lignocellulosic biomaterial provides an economical and renewable natural bioresource for the large-scale, gainful biofuel production, as well as the production of fine bulk chemicals, which may include industrial biocatalysts. To this end, the laccase-inducing aptitude of some agroindustrial
Sudipta Chatterjee et al.
Scientific reports, 9(1), 11658-11658 (2019-08-14)
A dual-responsive hydrogel (pH/temperature) was developed from a thermos-responsive polymer, pluronic F-127 (PF127), and pH-responsive polymers, N,N,N-trimethyl chitosan (TMC) and polyethylene glycolated hyaluronic acid (PEG-HA). Gallic acid, the principal component of the traditional Chinese drug Cortex Moutan was loaded into
Assessment of the joint effect of laccase and cellobiose dehydrogenase on the
decolouration of different synthetic dyes
Enayatzamir K
Journal of Hazardous Materials, 169, 176-181 (2009)
Renata M Lataro et al.
American journal of hypertension, 32(6), 579-587 (2019-03-16)
Acetylcholinesterase inhibition prevents autonomic imbalance, reduces inflammation, and attenuates the development of hypertension. Considering that vascular dysfunction is a crucial feature of arterial hypertension, we investigated the effects of chronic administration of acetylcholinesterase inhibitors-pyridostigmine or donepezil-on vascular reactivity of spontaneously

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service