Skip to Content
Merck
All Photos(1)

Key Documents

14610C

SAFC

EX-CELL® 610-HSF Serum-Free Medium for Hybridoma Cells

Low-protein, with L-glutamine, liquid, sterile-filtered, suitable for cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$69.00
1000 ML
$118.00

$69.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
500 ML
$69.00
1000 ML
$118.00

About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

$69.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

description

Drug Master File available
for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

storage condition

protect from light

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

components

glucose: 3.45 g/L
NaHCO3: 1.60 g/L
L-glutamine: 0.333 g/L
HEPES: 7.5 mM

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

EX-CELL 610-HSF is a low-protein (11 mg/L) serum-free medium. Originally developed to support the growth of hybrid cells in culture, it has been shown to support a wide range of cells including lymphoid and epithelial cells and B cell hybridomas of murine, rat and human origin. EX-CELL 610-HSF has been used in stationary culture systems and in large-scale bioreactors. In both types of culture, the production of cellular products, particularly monoclonal antibodies, has been shown to equal or exceed levels seen when the same cells are cultured in the presence of Fetal Bovine Serum (FBS).

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gunasekaran Dhandapani et al.
mAbs, 11(3), 583-592 (2019-01-09)
We introduce a new concept and potentially general platform for antibody (Ab) purification that does not rely on chromatography or specific ligands (e.g., Protein A); rather, it makes use of detergent aggregates capable of efficiently capturing Ab while rejecting hydrophilic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service