Skip to Content
Merck
All Photos(1)

Key Documents

137263

SAFC

Tromethamine

EMPROVE® EVOLVE

Pharma Manufacturing

Synonym(s):

Tromethamine, Tris

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11NO3
CAS Number:
Molecular Weight:
121.14
MDL number:
UNSPSC Code:
12352116
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

product line

EMPROVE® EVOLVE

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, microbial, and trace metals, tested

suitability

suitable for manufacturing use

storage temp.

2-30°C

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tromethamine is a commonly used biological buffer which is also referred to as Tris or Trizma. Tris as a buffer has very broad usage in diagnostics and downstream purifications. The pKa of Tris is 7.77 which corresponds closely to a physiological pH of 7.36.Tromethamine is used in biopharmaceutical buffer formulations (both upstream and downstream), electrophoresis separations and diagnostic reagent formulations. Tromethamine is especially useful in downstream purification processes.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service