13067C
Bovine Calf Serum
Australia origin, γ-irradiated by SER-TAIN™ process, sterile-filtered, suitable for cell culture
Synonym(s):
Calf Serum
About This Item
sterile-filtered
Recommended Products
biological source
bovine (calf)
sterility
γ-irradiated by SER-TAIN™ process
sterile-filtered
composition
Hemoglobin, ≤35 mg/dL
origin
Australia origin
technique(s)
cell culture | mammalian: suitable
impurities
≤50 EU/mL Endotoxin
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Application
Testing Method
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Find answers to questions (FAQs) about fetal bovine serum (FBS) that is gamma-irradiated by the SER-TAIN™ process. Request a sample of FBS at Sigma-Aldrich.com.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service