Skip to Content
Merck
All Photos(1)

Key Documents

10817

Sigma-Aldrich

Apolipoprotein A-I from human plasma

≥90% (GE)

Synonym(s):

Apo A-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human plasma

Assay

≥90% (GE)

form

lyophilized solid

mol wt

Mr ~28300

solubility

H2O: 1 mg/mL, clear, colorless

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... APOA1(335)

Looking for similar products? Visit Product Comparison Guide

General description

Apolipoprotein A-I (apoA-I) is the chief protein component of high density lipoproteins (HDL) and a major determinant of HDL structure, composition, and stability. It is an exchangeable apolipoprotein and plays pivotal role in the metabolism of cholesterol.

Application

Apolipoprotein A-I (apoA-I) was used for protecting the heart against vascular disease, it also protects other vascular beds and organs. It was protective against atherosclerosis through its ability to intervene in reverse cholesterol transport.

Biochem/physiol Actions

High-density lipoproteins are antiatherogenic and cardioprotective by facilitating reverse cholesterol transport (RCT). Some strategies for elevating apolipoprotein A-I levels are studied as therapies for atherosclerosis.

Preparation Note

lyophilized from 0.01 M ammonium hydrogen carbonate

Other Notes

A principal protein component of high-density lipoproteins. Protein - lipid interactions

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott M Gordon et al.
Current opinion in endocrinology, diabetes, and obesity, 19(2), 109-114 (2012-01-27)
To review recently published advances in the development of apolipoprotein A-I (apoA-I) mimetic peptides as a potential treatment for cardiovascular diseases. Various apoA-I mimetic peptides are currently in development and these display potent cardioprotective features that can rival or even
R.B. Gennis et al.
Annual Review of Biophysics and Biophysical Chemistry, 6, 195-195 (1977)
Jens O Lagerstedt et al.
Biochimica et biophysica acta, 1821(3), 448-455 (2012-01-17)
Apolipoprotein A-I (apoA-I) is the major protein component of high density lipoproteins (HDL) and plays a central role in cholesterol metabolism. The lipid-free/lipid-poor form of apoA-I is the preferred substrate for the ATP-binding cassette transporter A1 (ABCA1). The interaction of
Weiling Wang et al.
Journal of lipid research, 51(9), 2560-2570 (2010-05-26)
The relationship between high-density lipoprotein and pulmonary function is unclear. To determine mechanistic relationships we investigated the effects of genetic deletion of apolipoprotein A-I (apoA-I) on plasma lipids, paraoxonase (PON1), pro-inflammatory HDL (p-HDL), vasodilatation, airway hyperresponsiveness and pulmonary oxidative stress
Ryan J Henderson et al.
Lipids in health and disease, 8, 27-27 (2009-07-25)
Haptoglobin is a plasma protein that scavenges haemoglobin during haemolysis. Phospholipid Transfer Protein (PLTP) transfers lipids from Low Density Lipoproteins (LDL) to High Density Lipoproteins (HDL). PLTP is involved in the pathogenesis of atherosclerosis which causes coronary artery disease, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service