The growth rate of these cells is subject to multiple factors, including the age and origin of the cells, growth conditions, and media. While the doubling time of these cells typically falls within a range of 22 to 46 hours, it is also not uncommon to observe a variation of up to ±10 hours beyond this range.
102-05A
Human Epidermal Keratinocytes: HEK, adult
Synonym(s):
HEK cells
About This Item
Recommended Products
biological source
human neonatal foreskin or adult skin (normal)
Quality Level
packaging
pkg of 500,000 cells
manufacturer/tradename
Cell Applications, Inc
growth mode
Adherent
karyotype
2n = 46
morphology
Epidermal
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
allergies; cancer
shipped in
dry ice
storage temp.
−196°C
General description
Epidermal keratinocytes comprise 90 % of all cells found in the epidermis, the outermost layer of skin. They produce keratin, the intermediate filament, largely responsible for skin’s barrier function. Through their interactions with epidermal melanocytes, keratinocytes obtain melanin and store it as a means of protection from UV radiation.
HEK provide an excellent model system to study many aspects of epithelial function and disease, particularly those related to skin biology and toxicology, as well as HPV infection.
When grown on inserts and provided with the liquid/air interface, HEK can differentiate into a stratified squamous epithelium and serve as a more physiological 3D tissue model for in vitro studies.
Cell Line Origin
Application
Components
Preparation Note
- Primary culture, >500,000 cells in Basal Medium containing 10% FBS & 10% DMSO
- Can be cultured at least 16 doublings
Subculture Routine
Disclaimer
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Are there any modifications applied to the 102-05A cell line from primary cells? I am conducting experiments with this cell line, but the cells are growing much faster than I expected.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service