Skip to Content
Merck

08730

1,5-Diaminopentane dihydrochloride 1 M solution

Synonym(s):

1,5-Diaminopentane dihydrochloride solution, Additive Screening Solution 33/Kit-No 78374

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

aqueous solution

density

1.03 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

Cl.NCCCCCN

InChI

1S/C5H14N2.2ClH/c6-4-2-1-3-5-7;;/h1-7H2;2*1H

InChI key

FGNLEIGUMSBZQP-UHFFFAOYSA-N



Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library