Skip to Content
Merck

Skip To

08450

7-Amino-4-methyl-3-coumarinacetic acid N-succinimidyl ester

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Synonym(s):

7-(Dimethylamino)coumarin-4-acetic acid-NHS, 7-Amino-4-methyl-3coumarinylacetyl, N-Succinimidyl 7-amino-4-methyl-3-coumarinylacetate, AMCA-NHS, Succinimidyl-7-amino-4-methylcoumarin-3-acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
5 mg
Check Cart for Availability
$799.00

About This Item

Empirical Formula (Hill Notation):
C16H14N2O6
CAS Number:
Molecular Weight:
330.29
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352108
MDL number:
Beilstein/REAXYS Number:
9007679

$799.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Level

assay

≥90% (HPLC)

solubility

DMF: soluble, DMSO: soluble

fluorescence

λex 354 nm; λem 440 nm in methanol

suitability

suitable for fluorescence

storage temp.

−20°C

InChI

1S/C16H14N2O6/c1-8-10-3-2-9(17)6-12(10)23-16(22)11(8)7-15(21)24-18-13(19)4-5-14(18)20/h2-3,6H,4-5,7,17H2,1H3

InChI key

ILKQVTIMOGNSAS-UHFFFAOYSA-N

Application

AMCA-NHS has been utilized in the development of a donor europium chelate fluorochrome for use in LRET based analyses by first reacting with an amino group and then with DTPA anhydride . Utilized in FRET based studies, acting as the donor dye and can be paired with fluorescein-5-isothiocyanate as the acceptor dye . A blue fluorescent dye reactive towards primary amine groups on proteins, peptides, antibodies, and other biomolecules (at pH 7.0-9.0).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Labeling of lysyl residues in proteins[1]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
368015515603-32-1501
suitability

suitable for fluorescence

suitability

-

suitability

suitable for fluorescence

suitability

-

fluorescence

λex 354 nm; λem 440 nm in methanol

fluorescence

λex 445 nm; λem 482 nm in 0.1 M phosphate pH 7.0

fluorescence

λex 386 nm; λem 448 nm in 0.1 M Tris pH 9.0

fluorescence

-

solubility

DMF: soluble, DMSO: soluble

solubility

DMF: soluble, DMSO: soluble, acetonitrile: soluble

solubility

DMF: soluble, DMSO: soluble

solubility

5% acetic acid: 1 mg/mL, DMSO: 1 mg/mL, water: 1 mg/mL

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

200

assay

≥90% (HPLC)

assay

≥96.0% (HPLC)

assay

≥95.0% (coupling assay to aminopropyl silica gel)

assay

≥98% (HPLC)

product line

BioReagent

product line

BioReagent

product line

-

product line

-


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



H I Stefanova et al.
Biochemistry, 32(1), 356-362 (1993-01-12)
The (Ca(2+)-Mg2+)-ATPase of sarcoplasmic reticulum was labeled with the succinimidyl ester of 7-amino-4-methylcoumarin-3-acetic (AMCA). Although a large number of residues were labeled, it was found that Lys-492 was labeled preferentially at pH values between 6 and 8, consistent with an



Global Trade Item Number

SKUGTIN
234966-5G04061836683351
234966-25G04061836683344
08450-5MG04061833276686

Questions

Reviews

No rating value

Active Filters