Skip to Content
Merck
All Photos(1)

Key Documents

07211

Sigma-Aldrich

Streptavidin−Gold from Streptomyces avidinii

40 nm particle size

Synonym(s):

Streptavidin Gold conjugate

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$463.00

$463.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$463.00

About This Item

MDL number:
UNSPSC Code:
51102829
NACRES:
NA.25

$463.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

particle size

40 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

For detection of biotinylated compounds.
Streptavidin gold can be used to study biochemicals and reagents, fluorescent conjugates, fluorescent probes, labels, particles and stains, and streptavidin and avidin conjugates. Streptavidin gold has been used in a study to develop a simple, rapid, and sensitive method for visual detection of sequence-specific DNA. Streptavidin gold has also been used in a study to report the first instance of a novel application of mid-infrared chemical imaging (IRCI) for the fluorophore-free detection and identification of mycoplasma species.

Physical form

Material in 25 mM PBS pH 7.4 with 10% Glycerol, 0.1% BSA and 0.01% Sodium Azide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M M Mossoba et al.
Journal of microbiological methods, 86(3), 383-386 (2011-06-23)
A novel application of mid-infrared chemical imaging (IRCI) for the fluorophore-free detection and identification of mycoplasma species is reported for the first time. The PCR-amplified biotinylated targets hybridized to microarray probes were treated with streptavidin-gold nanoparticles followed by silver enhancement.
Yajing Niu et al.
Analytical chemistry, 83(19), 7500-7506 (2011-09-02)
A simple, rapid, and sensitive method for visual detection of sequence-specific DNA was developed using hairpin DNA as the recognition element and hydroxylamine-enlarged gold nanoparticles (Au-NPs) as the signal producing component. In the assay, we employed a hairpin DNA probe
Zhi-Qiang Shen et al.
Biosensors & bioelectronics, 26(7), 3376-3381 (2011-01-25)
In this paper, we describe a novel method for detecting Escherichia coli (E. coli) O157:H7 by using a quartz crystal microbalance (QCM) immunosensor based on beacon immunomagnetic nanoparticles (BIMPs), streptavidin-gold, and growth solution. E. coli O157-BIMPs were magnetic nanoparticles loaded

Questions

  1. What is the O.D. of item 07211-1ML Streptavidin−Gold from Streptomyces avidinii?

    1 answer
    1. The optical density is >320 at 530nm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service