Skip to Content
Merck
All Photos(1)

Key Documents

07082

Sigma-Aldrich

DABCYL-Glu-Arg-Nle-Phe-Leu-Ser-Phe-Pro-EDANS trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

N-[4-[[4-(Dimethylamino)phenyl]azo]benzoyl]-L-α-glutamyl-L-arginyl-L-norleucyl-L-phenylalanyl-L-leucyl-L-seryl-L-phenylalanyl-N-[2-[(5-sulfo-1-naphthalenyl)amino]ethyl]-L-prolinamide trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,310.00

$1,310.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 30, 2025



Select a Size

Change View
1 MG
$1,310.00

About This Item

Empirical Formula (Hill Notation):
C76H98N16O15S1 · xC2HF3O2
CAS Number:
Molecular Weight:
1507.75 (free base basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

$1,310.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 30, 2025


Assay

≥95% (HPLC)

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CCCC[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCC(O)=O)NC(=O)c1ccc(cc1)\N=N\c2ccc(cc2)N(C)C)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc4ccccc4)C(=O)N5CCC[C@H]5C(=O)NCCNc6cccc7c(cccc67)S(O)(=O)=O
OC(=O)C(F)(F)F.CCCC[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCC(O)=O)NC(=O)c1ccc(cc1)\N=N\c2ccc(cc2)N(C)C)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc4ccccc4)C(=O)N5CCC[C@H]5C(=O)NCCNc6cccc7c(cccc67)S(O)(=O)=O

InChI

1S/C76H98N16O15S.C2HF3O2/c1-6-7-24-57(83-68(97)58(26-16-39-81-76(77)78)84-70(99)59(37-38-66(94)95)82-67(96)50-29-31-51(32-30-50)89-90-52-33-35-53(36-34-52)91(4)5)69(98)86-61(44-48-18-10-8-11-19-48)72(101)85-60(43-47(2)3)71(100)88-63(46-93)73(102)87-62(45-49-20-12-9-13-21-49)75(104)92-42-17-27-64(92)74(103)80-41-40-79-56-25-14-23-55-54(56)22-15-28-65(55)108(105,106)107;3-2(4,5)1(6)7/h8-15,18-23,25,28-36,47,57-64,79,93H,6-7,16-17,24,26-27,37-46H2,1-5H3,(H,80,103)(H,82,96)(H,83,97)(H,84,99)(H,85,101)(H,86,98)(H,87,102)(H,88,100)(H,94,95)(H4,77,78,81)(H,105,106,107);(H,6,7)/b90-89+;/t57-,58-,59-,60-,61-,62-,63-,64-;/m0./s1

InChI key

NNUMUURWINZVDB-PZOCAEISSA-N

Biochem/physiol Actions

Peptide substrate for the assay of malaria aspartylproteinase, peptide sequence derived from the cleavage site present in hemoglobin, with norleucine as a substitution for methionine in order to avoid problems related to oxidation[1].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Jiang et al.
Antimicrobial agents and chemotherapy, 45(9), 2577-2584 (2001-08-15)
Malarial parasites rely on aspartic proteases called plasmepsins to digest hemoglobin during the intraerythrocytic stage. Plasmepsins from Plasmodium falciparum and Plasmodium vivax have been cloned and expressed for a variety of structural and enzymatic studies. Recombinant plasmepsins possess kinetic similarity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service