Skip to Content
Merck

04555

Trypanothione trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

N1,N8-Bis(glutathionyl)spermidine disulfide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H47N9O10S2 · xC2HF3O2
CAS Number:
Molecular Weight:
721.85 (free base basis)
UNSPSC Code:
12352204
NACRES:
NA.32
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3645155
Assay:
≥98% (HPLC)
Form:
powder or crystals
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder or crystals

solubility

water: soluble 1 g/L

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.N[C@@H](CCC(=O)N[C@H]1CSSCC(NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(=O)NCCCNCCCCNC(=O)CNC1=O)C(O)=O

InChI

1S/C27H47N9O10S2.C2HF3O2/c28-16(26(43)44)4-6-20(37)35-18-14-47-48-15-19(36-21(38)7-5-17(29)27(45)46)25(42)34-13-23(40)32-11-3-9-30-8-1-2-10-31-22(39)12-33-24(18)41;3-2(4,5)1(6)7/h16-19,30H,1-15,28-29H2,(H,31,39)(H,32,40)(H,33,41)(H,34,42)(H,35,37)(H,36,38)(H,43,44)(H,45,46);(H,6,7)/t16-,17-,18-,19?;/m0./s1

InChI key

BLTBWJNAZIKNTL-QZBRSAASSA-N

Biochem/physiol Actions

Metabolite in the glutathione metabolism. A novel bis(glutathionyl)spermidine cofactor for glutathione reductase in trypanosomatids.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library