Skip to Content
Merck
All Photos(1)

Key Documents

04482

Sigma-Aldrich

DEAE-Dextran

average mol wt 150,000

Synonym(s):

Diethylaminoethyl-Dextran

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$172.00

$172.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$172.00

About This Item

CAS Number:
UNSPSC Code:
12171500
NACRES:
NA.32

$172.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

mol wt

average mol wt 150,000

composition

nitrogen content, 2.5-4.5%

loss

≤10% loss on drying

storage temp.

−20°C

SMILES string

O(CCC)C(=O)c1ccc(cc1)C

InChI

1S/C11H14O2/c1-3-8-13-11(12)10-6-4-9(2)5-7-10/h4-7H,3,8H2,1-2H3

InChI key

LRNBWFXCDULFQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service