Skip to Content
Merck
All Photos(1)

Documents

03992

Sigma-Aldrich

Amiprofos methyl

PESTANAL®, analytical standard

Synonym(s):

O-Methyl O-(2-nitro-p-tolyl) N-isopropylphosphoramidothionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17N2O4PS
CAS Number:
Molecular Weight:
304.30
Beilstein:
2000833
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

grade

analytical standard
reference material

product line

PESTANAL®

Assay

≥98% (GC)

form

powder or crystals

shelf life

limited shelf life, expiry date on the label

mp

~65 °C

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(NC(C)C)Oc1ccc(C)cc1[N+]([O-])=O

InChI

1S/C11H17N2O4PS/c1-8(2)12-18(19,16-4)17-11-6-5-9(3)7-10(11)13(14)15/h5-8H,1-4H3,(H,12,19)

InChI key

VHEWQRWLIDWRMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Herbicide that prevents microtubule polymerization in plant cells, but not animal cells. Crop plants resistant to amiprofos-methyl and several other herbicides have been developed by insertion of a xenobiotic-detoxifying human gene CYP2B6.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G M Walker
Journal of general microbiology, 128(1), 61-71 (1982-01-01)
Of the seven antimicrotubular drugs tested, nocodazole, mebendazole and trifluralin at saturable concentrations failed to inhibit cell division in Schizosaccharomyces pombe, while carbendazim, thiabendazole and chloropropham each at 50 micrograms ml- and amiprophos methyl at 200 micrograms ml-1 completely arrested
F G Lux et al.
Genetics, 128(3), 549-561 (1991-07-01)
Through the isolation of suppressors of temperature-sensitive flagellar assembly mutations at the FLA10 locus of Chlamydomonas reinhardtii, we have identified six other genes involved in flagellar assembly. Mutations at these suppressor loci, termed SUF1-SUF6, display allele specificity with respect to
J Schwuchow et al.
Protoplasma, 159, 60-69 (1990-01-01)
Tip cells of dark-grown protonemata of the moss Ceratodon purpureus are negatively gravitropic (grow upward). They possess a unique longitudinal zonation: (1) a tip group of amylochloroplasts in the apical dome, (2) a plastid-free zone, (3) a zone of significant
S W James et al.
Genetics, 130(2), 305-314 (1992-02-01)
We previously described two types of genetic interactions among recessive mutations in the APM1 and APM2 loci of Chlamydomonas reinhardtii that may reflect a physical association of the gene products or their involvement in a common structure/process: (1) allele-specific synthetic
Harjinder Singh Sardar et al.
Plant physiology, 142(4), 1469-1479 (2006-10-24)
Arabinogalactan proteins (AGPs), a superfamily of plant hydroxyproline-rich glycoproteins, are present at cell surfaces. Although precise functions of AGPs remain elusive, they are widely implicated in plant growth and development. A well-characterized classical tomato (Lycopersicon esculentum) AGP containing a glycosylphosphatidylinositol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service