Skip to Content
Merck
All Photos(2)

Key Documents

A1952

Sigma-Aldrich

Ammonium sulfide solution

20% in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2S
CAS Number:
Molecular Weight:
68.14
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38030205
PubChem Substance ID:
NACRES:
NA.21
form:
liquid

form

liquid

Quality Level

concentration

20% in H2O
20.0-24.0% (titration by Na2S2O3)

storage temp.

room temp

SMILES string

N.N.S

InChI

1S/2H3N.H2S/h2*1H3;1H2

InChI key

UXXGWUQNRMPNKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium sulfide solution (aqueous ammonium sulfide) is a 20% solution in water. Ammonium sulfide is an inorganic salt with a sulfurous odor, which is commonly used in the food industry as a flavoring agent. The influence of ammonium sulfide treatment on (100)GaAs (gallium arsenide) surfaces has been studied by X-ray photoelectron spectroscopy (XPS) and reflection high-energy electron diffraction (RHEED). Its potential as an alternative to hydrogen sulfide for the transformation of activated amides to thioamides by thiolysis has been investigated. The reaction of ammonium sulfide with 3-hydroxy-2-butanone over a range of temperature has been reported to form various flavor compounds.

Application

Ammonium sulfide solution may be used to enhance the performance of type-II InAs/GaSb super lattice photodiodes by surface passivation technique.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Burdock GA.
Encyclopedia of Food and Color Additives, 1, 146-146 (1997)
Ammonium sulfide passivation of Type-II InAs/GaSb superlattice photodiodes.
Gin A, et al.
Applied Physics Letters, 84(12), 2037-2039 (2004)
Carpenter MS, et al.
J. Vac. Sci. Technol. B, 7(4), 845-850 (1989)
Mild method for the conversion of amides to thioamides.
Charette AB and Grenon M.
The Journal of Organic Chemistry, 68(14), 5792-5794 (2003)
Shuxia Xu et al.
Luminescence : the journal of biological and chemical luminescence, 25(4), 294-299 (2009-07-07)
In the present work, Fe(3)O(4)-carbon nanotubes (CNTs) composite was explored as a sensing material candidate for ammonium sulfide. Intense chemiluminescence emission can be observed during the catalytic oxidation of ammonium sulfide on the surface of Fe(3)O(4)-CNTs composite. Based on this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service