Skip to Content
Merck
All Photos(1)

Key Documents

676772

Sigma-Aldrich

Pyridine

ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
Beilstein:
103233
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
grade:
ACS reagent
Assay:
≥99.0%
bp:
115 °C (lit.)
vapor pressure:
10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

2.72 (vs air)

vapor pressure

10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

899 °F

expl. lim.

12.4 %

impurities

≤0.002% NH3
0.1% water

evapn. residue

≤0.002%

refractive index

n20/D 1.509 (lit.)

pH

8.5 (25 °C, 15.82 g/L)

bp

115 °C (lit.)

mp

−42 °C (lit.)

solubility

water: soluble

density

0.978 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.001%

cation traces

Cu: ≤5 ppm

SMILES string

C1=CN=CC=C1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridine (Pyr) is a nitrogen containing six membered heterocyclic compound commonly used as a base. It is a colorless liquid with a fishy odor. Pyridine moiety is present as a major component in a number of compounds like pyridine nucleotides,[1] pyridine alkaloids,[2] pyridine-based polymers[3] and pyridine-containing macrocycles.[4] It has wide application as a solvent, catalyst and as an intermediate for synthesis.[5] IR, Raman and UV spectra of pyridine have been analyzed and the values obtained were used to calculate its thermodynamic properties.[6] Photodegradation[7] and bacterial degradation of Pyr has been studied.[8]

Application

Pyridine may be used in the synthesis of the following:
  • complexes with cobalt(II) halides[9]
  • pyridine-d 5, a deuterated form of pyridine[10]
  • ionic liquid matrices (ILMs)[11]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Pyridine coenzymes of subcellular tissue fractions.
Jacobson KB and Kaplan NO.
The Journal of Biological Chemistry, 226(2), 603-613 (1957)
Nickel (II) and copper (II) complexes with pyridine-containing macrocycles bearing an aminopropyl pendant arm: synthesis, characterization, and modifications of the pendant amino group.
Herrera AM, et al.
Dalton Transactions, 5, 846-856 (2003)
Cyclostellettamines AF, pyridine alkaloids which inhibit binding of methyl quinuclidinyl benzilate (QNB) to muscarinic acetylcholine receptors, from the marine sponge, Stelletta maxima.
Fusetani N, et al.
Tetrahedron Letters, 35(23), 3967-3970 (1994)
The vibrational spectrum of pyridine and the thermodynamic properties of pyridine vapors.
Kline Jr CH and Turkevich J.
J. Chem. Phys. , 12(7), 300-309 (1944)
Bacterial degradation of pyridine, indole, quinoline, and their derivatives under different redox conditions.
Fetzner, S.
Applied Microbiology and Biotechnology, 49(3), 237-250 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service