Skip to Content
Merck
All Photos(1)

Key Documents

60109

Sigma-Aldrich

Potassium carbonate

puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$54.50
500 G
$98.10
1 KG
$184.00

$54.50

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
250 G
$54.50
500 G
$98.10
1 KG
$184.00

About This Item

Linear Formula:
K2CO3
CAS Number:
Molecular Weight:
138.21
Beilstein:
4267587
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
form:
powder or crystals

$54.50

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

ACS reagent
puriss. p.a.

Assay

≥99.0% (T)

form

powder or crystals

impurities

≤0.001% total nitrogen (N)
≤0.004% total sulfur (as SO4)

loss

≤1% loss on drying, 300 °C

pH

11-13 (25 °C, 138 g/L)

mp

891 °C (lit.)

anion traces

chloride (Cl-): ≤30 mg/kg
phosphate (PO43-): ≤10 mg/kg
silicate (as SiO2): ≤50 mg/kg

cation traces

Ag: ≤5 mg/kg
Al: ≤5 mg/kg
As: ≤0.5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Tl: ≤5 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[K+].[K+].[O-]C([O-])=O

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BWHMMNNQKKPAPP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium carbonate is an inorganic base.[1] It participates as a base in the palladacycle catalyzed Heck reaction of chlorobenzene with styrene.[2]

Application


  • Improved 2-pyridyl reductive homocoupling reaction using biorenewable solvent Cyrene (dihydrolevoglucosenone).: This study leverages potassium carbonate to enhance the homocoupling reaction of 2-pyridyl groups, showcasing its application in green chemistry for sustainable synthesis practices (Webb et al., 2023).

  • Synthesis, Characterization and Biological Evaluation of Magnolol and Honokiol Derivatives with 1,3,5-Triazine of Metformin Cyclization.: Potassium carbonate is used as a base in the synthesis of novel triazine derivatives, demonstrating its role in the development of biologically active compounds in pharmaceutical research (Ren et al., 2020).

  • Synthesis of (18) F-radiolabeled organophosphine fluorides for thiol-chemoselective peptide conjugation.: This article highlights the use of potassium carbonate in the preparation of radiolabeled compounds, essential for advancements in peptide conjugation and imaging techniques in molecular biology (Zhuang et al., 2020).

  • Molten-salt treatment of waste biomass for preparation of carbon with enhanced capacitive properties and electrocatalytic activity towards oxygen reduction.: Potassium carbonate is employed in molten-salt treatments to convert biomass into high-performance carbon materials, illustrating its importance in energy storage and environmental applications (Lu et al., 2016).

Other Notes

Reagent for the decomposition of silicates by fusioncitationcitation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Bock
Aufschlussmethoden der anorg. & org. Chemie (1972)
Z. Sulcek et al.
Methods of Decomposition in Inorganic Analysis (1989)
Robert Möckel et al.
Organic letters, 17(7), 1644-1647 (2015-03-21)
The cobalt-catalyzed Diels-Alder reaction of trimethylsilyl-substituted alkynes with 1,3-dienes led to dihydroaromatic intermediates which were transformed into iodobenzene derivatives. For this transformation, the dihydroaromatic intermediates had to be oxidized and the trimethylsilyl-substituted arene had to undergo a silicon-iodine exchange reaction.
Synthesis, kinetic observations and characteristics of polyarylene ether sulphones prepared via a potassium carbonate DMAC process.
Viswanathan R, et al.
Polymer, 25(12), 1827-1836 (1984)
Chelating N-heterocyclic carbene ligands in palladium-catalyzed Heck-type reactions.
Herrmann WA, et al.
Journal of Organometallic Chemistry, 557(1), 93-96 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service