Skip to Content
Merck
All Photos(2)

Documents

404985

Sigma-Aldrich

Titanium(IV) chloride solution

0.09 M in 20% HCl

Synonym(s):

Titanium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl4
CAS Number:
Molecular Weight:
189.68
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

form

liquid

Quality Level

reaction suitability

reagent type: catalyst
core: titanium

concentration

0.09 M in 20% HCl

density

1.099 g/mL at 25 °C

SMILES string

Cl[Ti](Cl)(Cl)Cl

InChI

1S/4ClH.Ti/h4*1H;/q;;;;+4/p-4

InChI key

XJDNKRIXUMDJCW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the detection of peroxides in ethyl ether. Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines.
Titanium(IV) chloride (TiCl4) solution may be used to synthesize nanosized titanium dioxide and nano/micro-sized titania powders.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of solution concentration on the hydrothermal preparation of titania crystallites.
Yanqing Z, et al.
Journal of Materials Chemistry, 11(5), 1547-1551 (2001)
Reagent Chemicals (1992)
Titanium tetrachloride promoted hydrolysis of cephalosporin tert-butyl esters.
Valencic M, et al.
Tetrahedron Letters, 39(12), 1625-1628 (1998)
Mechanism of enhanced performance of dye-sensitized solar cell based TiO2 films treated by titanium tetrachloride.
Long-Yue Z, et al.
Chinese Physics Letters, 21(9), 1835-1835 (2004)
Precipitation of nanosized titanium dioxide from aqueous titanium (IV) chloride solutions by neutralization with MgO.
Li Y and Demopoulos GP.
Hydrometallurgy, 90(1), 26-33 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service