Skip to Content
Merck
All Photos(3)

Key Documents

402982

Sigma-Aldrich

Lead(II) oxide

ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$102.00
1 KG
$344.00

$102.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
250 G
$102.00
1 KG
$344.00

About This Item

Linear Formula:
PbO
CAS Number:
Molecular Weight:
223.20
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NB.24
grade:
ACS reagent
form:
powder, crystals or chunks

$102.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

grade

ACS reagent

vapor pressure

10 mmHg ( 0 °C)

Assay

≥99.0%

form

powder, crystals or chunks

reaction suitability

reagent type: catalyst
core: lead

impurities

≤0.02% Insoluble matter in dilute HOAc

pH

8-9 (20 °C, 100 g/L)

mp

886 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%
nitrate (NO3-): ≤0.01%

cation traces

Ag: ≤5 ppm
Ca: ≤0.005%
Cu: ≤0.005%
Fe: ≤0.002%
K: ≤0.005%
Na: ≤0.02%

SMILES string

O=[PbH2]

InChI

1S/O.Pb

InChI key

YEXPOXQUZXUXJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lead (II) oxide is an oxide of lead with lead in +2 oxidation state. It is amphoteric in nature, capable of reacting with both acid and base.[1] It exists in two forms, litharge and massicot.[2] Lead (II) oxide is commonly used in making lead glass, lead white and red lead.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Kidney,Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clugston M and Flemming R.
Advanced Chemistry, 340-340 (2000)
Daintith J.
The facts on file dictionary of chemistry, 161-161 (1999)
Eagleson M
Concise Encyclopedia Chemistry, 590-590 (1994)
J Berashevich et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(7), 075803-075803 (2013-01-24)
The electronic properties of polycrystalline lead oxide consisting of a network of single-crystalline α-PbO platelets and the formation of native point defects in the α-PbO crystal lattice are studied using first-principles calculations. The results suggest that the polycrystalline nature of
Marco Panizza et al.
Chemosphere, 90(4), 1455-1460 (2012-10-03)
In this paper the electrocatalytic properties of Ti-Ru-Sn ternary oxide (TiRuSnO(2)), PbO(2) and boron-doped diamond (BDD) anodes have been compared for the electrochemical oxidation of a real landfill leachate from an old municipal solid waste landfill (average values of COD

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service