Skip to Content
Merck
All Photos(3)

Key Documents

383090

Sigma-Aldrich

Ammonium cerium(IV) sulfate dihydrate

ACS reagent, ≥94%

Synonym(s):

Ceric ammonium sulfate, Cerium(IV) ammonium sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$201.00
500 G
$656.00

$201.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$201.00
500 G
$656.00

About This Item

Linear Formula:
Ce(NH4)4(SO4)4 · 2H2O
CAS Number:
Molecular Weight:
632.55
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
grade:
ACS reagent
form:
powder or crystals

$201.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

grade

ACS reagent

Assay

≥94%

form

powder or crystals

reaction suitability

reagent type: catalyst
core: cerium

impurities

≤0.05% insol. dil. H2SO4

mp

130 °C (lit.)

anion traces

phosphate (PO43-): ≤0.03%

cation traces

Fe: ≤0.01%

SMILES string

N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Ce.4H3N.4H2O4S.2H2O/c;;;;;4*1-5(2,3)4;;/h;4*1H3;4*(H2,1,2,3,4);2*1H2/q+4;;;;;;;;;;/p-4

InChI key

VCNAMBGKEDPVGQ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium cerium(IV) sulfate dehydrate is a cerium salt widely used in the volumetric analysis.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self-Poled Transparent and Flexible UV Light-Emitting Cerium Complex-PVDF Composite: A High-Performance Nanogenerator.
Garain S, et al.
ACS Applied Materials & Interfaces, 7(2), 1298-1307 (2015)
David CA and Hellier DG
Dictionary of Environmentally Important Chemicals, 92-92 (1997)
Analysis of phenolic compounds in health care products by low-pressure liquid-chromatography with monolithic column and chemiluminescent detection.
Ballesta-Claver J, et al.
Luminescence, 26(1), 44-53 (2011)
Cerium-initiated grafting of acrylonitrile onto cellulosic materials.
Hebeish A and Mehta PC.
Journal of Applied Polymer Science, 12(7), 1625-1647 (1968)
K H Gordon et al.
Organic letters, 3(1), 53-56 (2001-06-30)
[figure: see text] A novel benzyloxyaniline linker that uses ceric ammonium nitrate (CAN) as a cleavage reagent is described. Its application in the solid-phase synthesis of N-unsubstituted beta-lactams and secondary amides furnishes compounds in moderate to excellent yield (45-91%) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service