Skip to Content
Merck
All Photos(1)

Key Documents

363642

Sigma-Aldrich

Sodium periodate

10 wt. %, matrix silica gel support

Synonym(s):

Sodium (meta)periodate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaIO4
CAS Number:
Molecular Weight:
213.89
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

powder

reaction suitability

reagent type: oxidant

concentration

10 wt. %
9.0-11.0% (by Na2S2O3, titration)

matrix

silica gel support

pH

3.5-5.5 (25 °C, 107 g/L)

mp

300 °C (dec.) (lit.)

SMILES string

[Na+].[O-]I(=O)(=O)=O

InChI

1S/HIO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

JQWHASGSAFIOCM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium periodate (sodium (meta)periodate) supported on silica gel is a solid supported reagent.[1] It may be synthesized by adding silica gel to hot aqueous solution of sodium metaperiodate and evaporating to dryness followed by vigorous swirling and shaking.[2]

Application

Sodium periodate (sodium (meta)periodate) supported on silica gel may be used in the following reactions:
  • Oxidative cleavage of glycols to alkenes[3] and 1,2-diols to aldehydes.[4]
  • Phthalimide N-oxyl (PINO) radical generation from N-hydroxyphthalimide (NHPI).[5]
  • Selective oxidation of sulfides by microwave thermolysis process.[6]
  • Regeneration of ketones from ketoximes by microwave irradiation under solvent-free conditions.[7]
  • Preparation of 2,3-O-isopropylidene-D-glyceraldehyde.[2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1C - STOT RE 1

Target Organs

thymus gland

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid state regeneration of ketones from oximes on wet silica supported sodium periodate using microwaves.
Varma RS, et al.
Tetrahedron Letters, 38(51), 8819-8820 (1997)
A general procedure for a one-pot oxidative cleavage/Wittig reaction of glycols.
Dunlap NK, et al.
Tetrahedron Letters, 43(21), 3923-3925 (2002)
Selective oxidation of sulfides to sulfoxides and sulfones by microwave thermolysis on wet silica-supported sodium periodate.
Varma RS, et al
Tetrahedron Letters, 38(37), 6525-6528 (1997)
A New and Efficient Heterogeneous system for the oxidative cleavage of 1, 2-Diols and the Oxidation of Hydroquinones.
Daumas M, et al.
Synthesis, 1, 64-65 (1989)
A New and Efficient Heterogeneous System for the Phthalimide N-Oxyl (PINO) Radical Generation.
Coseri S.
European Journal of Organic Chemistry, 2007(11), 1725-1729 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service