Skip to Content
Merck
All Photos(2)

Documents

334448

Sigma-Aldrich

Molecular sieves

powder, Catalyst support, sodium Y zeolite

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23201100
eCl@ss:
32110204
NACRES:
SB.52

form

powder

particle size distribution

100 mesh (>=90%)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A molecular dynamics study of xenon sorbed in sodium Y zeolite. 1. Temperature and concentration dependence.
Santikary P, et al.
The Journal of Physical Chemistry, 96(25), 10469-10477 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service