Skip to Content
Merck
All Photos(1)

Key Documents

33214

Sigma-Aldrich

Acetic anhydride

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99% (GC)

Synonym(s):

Acetanhydride, Acetic acid anhydride, Acetyl acetate, Acetyl anhydride, Ethanoic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO)2O
CAS Number:
Molecular Weight:
102.09
Beilstein:
385737
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

3.5 (vs air)

vapor pressure

10 mmHg ( 36 °C)
4 mmHg ( 20 °C)

Assay

≥99% (GC)

autoignition temp.

629 °F

expl. lim.

10.3 %

impurities

≤0.003% non-volatile matter
≤0.01% KMnO4 red. matter (as O)

refractive index

n20/D 1.390 (lit.)

bp

138-140 °C (lit.)

mp

−73 °C (lit.)

density

1.08 g/mL (lit.)

anion traces

chloride (Cl-): ≤2 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤5 mg/kg

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.05 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.2 mg/kg

suitability

passes test for reaction against H2SO4

SMILES string

CC(=O)OC(C)=O

InChI

1S/C4H6O3/c1-3(5)7-4(2)6/h1-2H3

InChI key

WFDIJRYMOXRFFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Acetic anhydride is a colorless liquid. On dissolution in water, it undergoes solvolysis to afford acetic acid. It is widely employed as acetylation reagent for the acetylation of various alcohols. Wacher process (starting reagent + a ketene) and Knapsack process (starting reagent = acetaldehyde) have been reported for the industrial preparation of acetic anhydride. It is employed as solvent for various organic reactions.

Application

  • Modification of Quinoa Starches: Acetic anhydride is used to modify the structure of starch molecules, discussing its implications on improving bread quality and food processing (Wang et al., 2024).

Other Notes

The article number 33214-4X2.5L will be discontinued. Please order the single bottle 33214-2.5L which is physically identical with the same exact specifications.
The article number 33214-4X2.5L-M will be discontinued. Please order the single bottle 33214-2.5L-M which is physically identical with the same exact specifications.
The article number 33214-6X1L will be discontinued. Please order the single bottle 33214-1L which is physically identical with the same exact specifications.
The article number 33214-6X1L-M will be discontinued. Please order the single bottle 33214-1L-M which is physically identical with the same exact specifications.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Barath et al.
The American journal of pathology, 137(3), 503-509 (1990-09-01)
We used immunohistochemistry to detect tumor necrosis factor (TNF) and in situ hybridization to detect TNF messenger RNA (mRNA) in the intimal mesenchymal-appearing cells and in the medial smooth muscle cells of human atherosclerotic arteries. Medial smooth muscle cells showed
Acetylation of anisole by acetic anhydride over a HBEA zeolite-Origin of deactivation of the catalyst.
Rohan D, et al.
J. Catal., 177(2), 296-305 (1998)
David G Simmons et al.
BMC genomics, 9, 352-352 (2008-07-30)
The Prolactin (PRL) hormone gene family shows considerable variation among placental mammals. Whereas there is a single PRL gene in humans that is expressed by the pituitary, there are an additional 22 genes in mice including the placental lactogens (PL)
Francesca Morello et al.
Cell reports, 33(2), 108268-108268 (2020-10-15)
Tegmental nuclei in the ventral midbrain and anterior hindbrain control motivated behavior, mood, memory, and movement. These nuclei contain inhibitory GABAergic and excitatory glutamatergic neurons, whose molecular diversity and development remain largely unraveled. Many tegmental neurons originate in the embryonic
Eagleson M.
Concise Encyclopedia Chemistry, 5-5 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service