Skip to Content
Merck
All Photos(1)

Key Documents

31128

Sigma-Aldrich

Barium nitrate

puriss. p.a., ACS reagent, ≥99%, ≥52% Ba basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(NO3)2
CAS Number:
Molecular Weight:
261.34
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Assay:
≥99%
form:
solid

grade

ACS reagent
puriss. p.a.

Assay

≥99%

form

solid

concentration

≥52% Ba

impurities

≤0.001% ammonium (NH4)

pH

5-7 (20 °C, 5%)

mp

592 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg

cation traces

Ca: ≤20 mg/kg
Fe: ≤2 mg/kg
K: ≤50 mg/kg
Mg: ≤20 mg/kg
Na: ≤50 mg/kg
Sr: ≤500 mg/kg
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ba.2NO3/c;2*2-1(3)4/q+2;2*-1

InChI key

IWOUKMZUPDVPGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Suicidal poisoning with barium nitrate].
Z Kołaciński
Polski tygodnik lekarski (Warsaw, Poland : 1960), 42(11), 339-341 (1987-03-16)
M Monaco et al.
La Medicina del lavoro, 82(5), 439-445 (1991-09-01)
Barium nitrate, which is used in industry in the production of green signal lights, to remove gases from vacuum tubes, and in the production of barium oxide, was assayed to assess the possible mutagenic effects using both the Ames test
[Comparative toxicity of various barium compounds].
T A Akinfieva et al.
Gigiena truda i professional'nye zabolevaniia, (6)(6), 45-46 (1984-06-01)
G G Liu et al.
Journal of environmental sciences (China), 13(1), 75-78 (2001-10-10)
In this paper, the effect of Ba(NO3)2 on the efficiency of sulfur fixation of calcium oxide during coal combustion was studied. The results showed that addition of barium nitrate to the CaO can enhance the sulfur removal rate of CaO
Porous YSZ impregnated with La0.4Sr0.5Ba0.1TiO3 as a possible composite anode for SOFCs fueled with sour feeds.
Vincent AL, et al.
Journal of Power Sources, 215, 301-306 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service