Skip to Content
Merck
All Photos(1)

Key Documents

270318

Supelco

Benzonitrile

suitable for HPLC, 99.9%

Synonym(s):

Phenyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CN
CAS Number:
Molecular Weight:
103.12
Beilstein:
506893
EC Number:
MDL number:
UNSPSC Code:
12190000
eCl@ss:
39031505
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
191 °C (lit.)
Pricing and availability is not currently available.

grade

HPLC grade

Assay

99.9%

form

liquid

purified by

glass distillation

expl. lim.

0.34-6.3 %

technique(s)

HPLC: suitable

impurities

≤0.03% water

evapn. residue

<0.0005%

refractive index

n20/D 1.528 (lit.)

bp

191 °C (lit.)

mp

−13 °C (lit.)

λ

H2O reference

UV absorption

λ: 300 nm Amax: 1.0
λ: 310 nm Amax: 0.40
λ: 335 nm Amax: 0.03
λ: 360-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

N#Cc1ccccc1

InChI

1S/C7H5N/c8-6-7-4-2-1-3-5-7/h1-5H

InChI key

JFDZBHWFFUWGJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzonitrile and acetonitrile complexes of ruthenium ammines.
Clarke RE and Ford PC.
Inorganic Chemistry, 9(2), 227-235 (1970)
Lanthanide-imido complexes and their reactions with benzonitrile.
Dongmei Cui et al.
Angewandte Chemie (International ed. in English), 44(6), 959-962 (2004-12-22)
A multiphase reaction medium including pressurized carbon dioxide and water for selective hydrogenation of benzonitrile with a Pd/Al2O3 catalyst.
Yoshida H, et al.
Applied Catalysis A: General, 456, 215-222 (2013)
Yoshimitsu Hashimoto et al.
Organic & biomolecular chemistry, 10(30), 6003-6009 (2012-05-19)
A variety of highly functionalized polycyclic isoxazoles are prepared by a two-step protocol: (1) 1,3-dipolar cycloaddition of o,o'-disubstituted benzonitrile oxides to para-quinone mono-acetals, then (2) dehydrogenation. The cycloaddition proceeds in a regioselective manner, favouring the formation of the 4-acyl cycloadducts
Jessica L Durham et al.
Inorganic chemistry, 51(14), 7825-7836 (2012-07-07)
The preparation of two new families of hexanuclear rhenium cluster complexes containing benzonitrile and phenyl-substituted tetrazolate ligands is described. Specifically, we report the preparation of a series of cluster complexes with the formula [Re(6)Se(8)(PEt(3))(5)L](2+) where L = benzonitrile, p-aminobenzonitrile, p-methoxybenzonitrile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service